Learn More
Eutech™ KCl Calibration Solution
Click to view available options
Quantity:
60 mL
480 mL
Conductivity:
100 μS/cm
111.8 mS/cm
12.88 mS/cm
1413 μS/cm
2764 μS/cm
500 μS/cm
84 μS/cm
Unit Size:
480mL
500mL
60mL
Description
Eutech™ KCl Calibration Solutions are used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
- Packed in 60mL or 480 ml durable plastic bottles
- Economical and convenient to use
Specifications
Specifications
| Description | 12.88 mS/cm KCl Calibration Solution |
| Conductivity | 12.88 mS/cm |
| For Use With (Equipment) | Conductivity Meters and Electrodes |
| Solution Type | Calibration Solution |
| Quantity | 60 mL |