missing translation for 'onlineSavingsMsg'
Learn More

Abcam EUK 134, SOD/Catalase Mimetic, >98% Purity, Solid, 5 mg

Product Code. 32109076 Shop All Abcam Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32109076 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32109076 Supplier Abcam Supplier No. ab1455935mg
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Synthetic SOD/catalase mimetic with neuroprotective and anti-ischemic effects.

  • Molecular Weight: 416.7 Da
  • Purity: >98%
  • Form: Solid
  • Function: SOD/catalase mimetic
  • Activity: Consumes H2O2 with an initial reaction rate of 234 μM/min
  • Biological Effects: Ameliorates nNOSμ translocation and skeletal muscle fiber atrophy during short-term mechanical unloading; shows neuroprotective and anti-ischemic effects in vivo
  • Storage: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Antioxidant
Buffer 100% manganese(2+);2-methoxy-6-[2-[(3-methoxy-2-oxidophenyl)methylideneamino]ethyliminomethyl]phenolate;chloride
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrate H2O2
Target Molecule H2O2
Formulation 100% manganese(2+);2-methoxy-6-[2-[(3-methoxy-2-oxidophenyl)methylideneamino]ethyliminomethyl]phenolate;chloride
Quantity 5 mg
For Use With (Application) Neuroprotection, Anti-ischemia, Skeletal muscle fiber atrophy research
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.