missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ETV2 Monoclonal Antibody (827940)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561859
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Binds to DNA sequences containing the consensus pentanucleotide 5'-CGGA[AT]-3'.
Specifications
| ETV2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P41163 | |
| ETV2 | |
| E. coli-derived recombinant mouse ETV2/ER71, Asn125-Arg231. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 827940 | |
| Unconjugated | |
| ETV2 | |
| ER71; ets related protein 71; ETS translocation variant 2; ETS variant 2; ets variant gene 2; ETS variant transcription factor 2; ets-related protein 71; Etsrp71; ETV2 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 14008 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction