missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ETV1 Antibody (CL15257) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43654-25ul
This item is not returnable.
View return policy
Description
ETV1 Monoclonal antibody specifically detects ETV1 in Human samples. It is validated for Western Blot
Specifications
| ETV1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL | |
| DKFZp781L0674, ER81ets variant gene 1, ETS translocation variant 1, ets variant 1, Ets-related protein 81, MGC104699, MGC120533, MGC120534 | |
| This antibody was generated using a synthetic peptide of P50549, with the exact immunogen sequence remaining proprietary. | |
| 25 μL | |
| Stem Cell Markers | |
| 2115 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot | |
| CL15257 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction