missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etofenamate Palmitate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Etofenamate Palmitate
Specifications
Specifications
| Chemical Name or Material | Etofenamate Palmitate |
| CAS | 81427-95-4 |
| Analyte or Component Names | Etofenamate Palmitate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C34 H48 F3 N O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Etofenamate Palmitate |
| InChI Formula | InChI=1S/C34H48F3NO5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-22-32(39)42-25-23-41-24-26-43-33(40)30-20-15-16-21-31(30)38-29-19-17-18-28(27-29)34(35,36)37/h15-21,27,38H,2-14,22-26H2,1H3 |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCCOCCOC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F |
| IUPAC Name | 2-(2-hexadecanoyloxyethoxy)ethyl 2-[3-(trifluoromethyl)anilino]benzoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction