missing translation for 'onlineSavingsMsg'
Learn More

Etofenamate Myristate, Mikromol™

Product Code. 30050122
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30050122

Brand: Mikromol MM0105.08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Etofenamate Myristate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Etofenamate Myristate
CAS 81427-99-8
Analyte or Component Names Etofenamate Myristate
Shipping Condition Room Temperature
Molecular Formula C32 H44 F3 N O5
Certifications/Compliance ISO 17025
Synonym Etofenamate Myristate
InChI Formula InChI=1S/C32H44F3NO5/c1-2-3-4-5-6-7-8-9-10-11-12-20-30(37)40-23-21-39-22-24-41-31(38)28-18-13-14-19-29(28)36-27-17-15-16-26(25-27)32(33,34)35/h13-19,25,36H,2-12,20-24H2,1H3
SMILES CCCCCCCCCCCCCC(=O)OCCOCCOC(=O)c1ccccc1Nc2cccc(c2)C(F)(F)F
IUPAC Name 2-(2-tetradecanoyloxyethoxy)ethyl 2-[3-(trifluoromethyl)anilino]benzoate
Molecular Weight (g/mol) 579.69
Quantity 100 mg
Formula Weight 579.3172 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.