missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Di Na Salt Dihydr. (Crystalline Powd./Electrophor.), Fisher BioReagents™

Product Code. p-25811
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 10618973

Brand: Fisher BioReagents BP120500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For minimization of metal ion impurities in reaction buffers

The disodium salt of EDTA is a chelating agent for metal ions and is used extensively in molecular biology to minimize metal ion impurities in reaction buffers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
IUPAC Name disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Decomposition Information >252°C
Melting Point 252°C
Color White
pH 4 to 6
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2Na)2·2H2O
Quantity 500 g
Merck Index 15, 3565
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 372.23
Percent Purity ≥99.0%
Grade Electrophoresis
Physical Form Solid
Chemical Name or Material Ethylenediamine Tetraacetic Acid, Disodium Salt Dihydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, disodium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.