Learn More
Ethyl trans-2-hexenoate, 98%, Thermo Scientific™
Product Code. 11482108
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482108 50 g 50g
11472108 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11482108 Supplier Thermo Scientific Alfa Aesar Supplier No. A15136.18
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27829-72-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00036541
InChI Key SJRXWMQZUAOMRJ-VOTSOKGWSA-N
Synonym ethyl hex-2-enoate, ethyl 2-hexenoate, ethyl trans-2-hexenoate, 2-hexenoic acid, ethyl ester, ethyl e-hex-2-enoate, ethyl e-2-hexenoate, ethyl 2e-hexenoate, 2-hexenoic acid, ethyl ester, 2e, unii-ol4euz6c13, 2-hexenoic acid, ethyl ester, e
PubChem CID 5364778
IUPAC Name ethyl (E)-hex-2-enoate
SMILES CCCC=CC(=O)OCC
Specifications
Density 0.95
Boiling Point 62°C (25 mmHg)
Flash Point 62°C (143°F)
Refractive Index 1.434
Quantity 50 g
UN Number UN3272
Beilstein 1701323
Formula Weight 142.2
Percent Purity 98%
Chemical Name or Material Ethyl trans-2-hexenoate