Learn More
Ethyl cinnamate, 98% trans
Product Code. 10615012 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
Product Code. 10615012
Brand: Thermo Scientific Acros undefined
Request Bulk or Custom Format
CAS: 103-36-6 | C11H12O2 | 176.21 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 103-36-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.21
InChI Key KBEBGUQPQBELIU-CMDGGOBGSA-N
Synonym ethyl cinnamate, ethyl 3-phenylacrylate, ethylcinnamate, cinnamic acid ethyl ester, ethylcinnamoate, cinnamic acid, ethyl ester, ethyl trans-cinnamate, e-ethyl cinnamate, ethyl 3-phenyl-2-propenoate, ethyl benzylideneacetate
PubChem CID 637758
ChEBI CHEBI:4895
IUPAC Name ethyl (E)-3-phenylprop-2-enoate
SMILES CCOC(=O)C=CC1=CC=CC=C1
Specifications
Acidity 1mg KOH/g max.
Melting Point 6.5°C to 7.5°C
Color Colorless to Yellow
Density 1.0460g/mL
Boiling Point 271.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH=CHCO2C2H5
Refractive Index 1.5580 to 1.5610
Quantity 500 g
Merck Index 15,2299
Specific Gravity 1.046
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 176.21
Percent Purity 98% trans
Physical Form Liquid
Chemical Name or Material Ethyl cinnamate