Learn More
Ethyl 4,4,4-trifluorocrotonate, 98%
Product Code. 11338345
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338345 5 g 5g
11328345 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11338345 Supplier Thermo Scientific Alfa Aesar Supplier No. L10285.06
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
CAS: 25597-16-4 | C6H7F3O2 | 168.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25597-16-4
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.12
MDL Number MFCD00009903
InChI Key ZKRJCMKLCDWROR-ONEGZZNKSA-N
Synonym ethyl 4,4,4-trifluorocrotonate, ethyl 2e-4,4,4-trifluorobut-2-enoate, ethyl4,4,4-trifluorocrotonate, e-ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluorocrotonic acid ethyl ester, ethyl e-4,4,4-trifluorobut-2-enoate, ethyl-4,4,4-trifluorocrotonate, ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluoro-but-2-enoic acid ethyl ester, 2-butenoic acid, 4,4,4-trifluoro-, ethyl ester, 2e
PubChem CID 5371261
IUPAC Name ethyl (E)-4,4,4-trifluorobut-2-enoate
SMILES CCOC(=O)\C=C\C(F)(F)F
Specifications
Density 1.125
Boiling Point 113°C to 115°C
Flash Point 25°C (77°F)
Refractive Index 1.3601
Quantity 5 g
UN Number UN3272
Beilstein 1707262
Formula Weight 168.12
Percent Purity 98%
Chemical Name or Material Ethyl 4,4,4-trifluorocrotonate