missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ESTRIOL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43293
This item is not returnable.
View return policy
Description
ESTRIOL Polyclonal antibody specifically detects ESTRIOL in Nonspecies specific samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
Specifications
| ESTRIOL | |
| Polyclonal | |
| Unconjugated | |
| PBS, pH 7.4, 50% glycerol | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Non-species specific | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence | |
| 0.5 mg/mL | |
| ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunohistochemistry-Frozen 5-20 μg/mL | |
| C18H24O3;Estratriol;Estriol;Oestriol;Ovestin;Ovestrion | |
| OVA - Estriol | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction