Learn More
Estramustine, Mikromol™
Product Code. 18031540
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18031540 25 mg 25mg
18013330 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18031540 Supplier Mikromol Supplier No. MM0100.010025
 Restricted Item
Estimated Shipment: 02-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Estramustine
TRUSTED_SUSTAINABILITY
Spezifikation
Chemical Name or Material Estramustine
CAS 2998-57-4
Analyte or Component Names Estramustine
Shipping Condition Room Temperature
Molecular Formula C23 H31 Cl2 N O3
Certifications/Compliance ISO 17025
Synonym Estradiol 3-[N,N-Bis(2-chloroethyl)carbamate], Estramustine
InChI Formula InChI=1S/C23H31Cl2NO3/c1-23-9-8-18-17-5-3-16(29-22(28)26(12-10-24)13-11-25)14-15(17)2-4-19(18)20(23)6-7-21(23)27/h3,5,14,18-21,27H,2,4,6-13H2,1H3/t18-,19-,20+,21+,23+/m1/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(OC(=O)N(CCCl)CCCl)ccc34)[C@@H]1CC[C@@H]2O
IUPAC Name [(8R,9S,13S,14S,17S)-17-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] N,N-bis(2-chloroethyl)carbamate
Molecular Weight (g/mol) 440.4
Quantity 25 mg
Formula Weight 439.1681 g/mol
Mehr anzeigen Weniger anzeigen