missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estradiol Antibody (RM343), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-77405
This item is not returnable.
View return policy
Description
Estradiol Monoclonal antibody specifically detects Estradiol in Nonspecies specific samples. It is validated for ELISA, Immunoassay, Competitive ELISA
Specifications
| Estradiol | |
| Monoclonal | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Non-species specific | |
| Purified |
| ELISA, Immunoassay, Competition Assay | |
| RM343 | |
| ELISA, Immunoassay, Competitive ELISA 0.5 - 1.5 ng/well on a 96-well plate pre-coated with a goat anti-rabbit IgG polyclonal antibody | |
| 17-Beta-Estradiol;17-beta-HSD | |
| Estradiol BSA Conjugate | |
| 100 μg | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction