Learn More
ESRP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23117-1-AP-150UL
This item is not returnable.
View return policy
Description
RBM35A, also known as ESRP2, is a mRNA splicing factor that with its related protein RBM35A (ESRP1) are coordinators of an epithelial cell-type-specific splicing program. Both RBM35B and RBM35A are involved in posttranscriptional regulation of a number of genes such as FGFR2, CD44, CTNND1, and ENAH by exerting a differential effect on protein translation via 5′ UTRs of mRNAs, suggesting that these proteins are global regulators of an epithelial regulatory network. Loss of this global ESRP-regulated epithelial splicing program induces the phenotypic changes in cell morphology that are observed during the epithelial-mesenchymal transition.Specifications
| ESRP2 | |
| Polyclonal | |
| Unconjugated | |
| ESRP2 | |
| ESRP2, RBM35B, RNA binding motif protein 35B, RNA binding protein 35B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 77411, 80004 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8K0G8, Q9H6T0 | |
| Esrp2 | |
| ESRP2 Fusion Protein Ag19436 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |