Learn More
Thermo Scientific™ ES Series Lab Refrigerators
Product Code. 16617892 Shop All Thermo Scientific Products
Click to view available options
Capacity (Metric):
158 L
Plug Type:
EU
Unit Size:
Each
Product Code. 16617892
Brand: Thermo Scientific™ 158RAEVTS
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Save valuable space with Thermo Scientific™ ES Series Lab Refrigerators, designed for general-purpose lab storage.
Features:
  • Integrated controller
  • Digital display
  • High- and low-temperature alarms
  • Standard door locks
  • Low energy consumption
  • Access ports
  • CE marked
Ordering Information:
Available in 50Hz regions only; not available in North America or other 60Hz regions
Recommended for:
General-purpose lab storage
  • Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Type General Purpose Refrigerator
Capacity (English) 5.5 cu. ft.
Capacity (Metric) 158 L
Temperature Range 1°C to 11°C
Refrigerant R600a
Door Style Hinged
Shelves 3
Cabinet Material Scratch-resistant Painted Steel
Insulation HCFC Free, CFC Free
Certifications/Compliance CE
Style Undercounter
Control Type Digital
Ports 1, 16 mm Side-Access Port
Defrost Automatic
Monitoring Options Alarm
Depth (Metric) Exterior 65.8 cm
Depth (Metric) Interior 48.0 cm
Height (Metric) Exterior 85.0 cm
Height (Metric) Interior 71.8 cm
Width (Metric) Exterior 59.9 cm
Width (Metric) Interior 50.0 cm
Plug Type EU
Voltage 230 V
Electrical Requirements 230 V, 50Hz
Frequency 50 Hz
Shipping Weight (Metric) 37.8 kg
Quantity Each