missing translation for 'onlineSavingsMsg'
Learn More

Erythropoietin R Rabbit anti-Human, Clone: 16, Novus Biologicals™

Rabbit Monoclonal Antibody

384.00€ - 571.00€

Specifications

Antigen Erythropoietin R
Clone 16
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18241096
View Documents
Novus Biologicals
NBP2-89615-50UL
50 μg
384.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18283746
View Documents
Novus Biologicals
NBP2-89615-100UL
100 μg
571.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Erythropoietin R Monoclonal specifically detects Erythropoietin R in Human samples. It is validated for Western Blot.
Specifications

Specifications

Erythropoietin R
Western Blot 1:500-1:2000
Monoclonal
Purified
RUO
Human
EPO-R, erythropoietin receptor, MGC138358
Recombinant Human Erythropoietin R protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
16
Western Blot
Unconjugated
Rabbit
Angiogenesis, Cancer, Immunology, Phospho Specific, Signal Transduction
0.2 um filtered solution in PBS
2057
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.