missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB4/Her4 Mouse anti-Human, Mouse, Clone: HFR-1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07160-20UG
This item is not returnable.
View return policy
Description
ErbB4/Her4 Monoclonal specifically detects ErbB4/Her4 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
| ErbB4/Her4 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
| avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
| Recombinant human ErbB4/Her4 protein fragment (around aa 1116-1269) (exact sequence is proprietary) | |
| 20 μg | |
| Angiogenesis, Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| 2066 | |
| Store at 4°C. | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence | |
| HFR-1 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion