missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB3/Her3 Antibody (6F6), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | ErbB3/Her3 |
|---|---|
| Clone | 6F6 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043186
|
Novus Biologicals
NBP3-26599-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043922
|
Novus Biologicals
NBP3-26599-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
| ErbB3/Her3 | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| A synthesized peptide derived from Human ErbB3/Her3 [UniProt P21860] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 6F6 | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2065 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title