missing translation for 'onlineSavingsMsg'
Learn More

F5, Horse, Polyclonal Antibody, Abnova™

Product Code. 16052627
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16052627 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16052627 Supplier Abnova Supplier No. PAB17092.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Horse polyclonal antibody raised against F5.

This gene encodes an essential cofactor of the blood coagulation cascade. This factor circulates in plasma, and is converted to the active form by the release of the activation peptide by thrombin during coagulation. This generates a heavy chain and a light chain which are held together by calcium ions. The activated protein is a cofactor that participates with activated coagulation factor X to activate prothrombin to thrombin. Defects in this gene result in either an autosomal recessive hemorrhagic diathesis or an autosomal dominant form of thrombophilia, which is known as activated protein C resistance. [provided by RefSeq

Specifications

Antigen F5
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Horse polyclonal antibody raised against F5.
Dilution ELISA (10 ug/mL) Western Blot (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene F5
Gene Alias FVL/PCCF
Gene Symbols F5
Host Species Equine
Immunogen Purified human F5.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2153
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.