Learn More
Eppendorf™ S-4-72 Rotor Adapters
Product Code. 15108364 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
1 x 250 mL Tubes
12 x 7/17 mL Tubes
13 x 5.5/12 mL Tubes
14 x 2.6/8 mL Tubes
2 x 50 mL Conical Tubes
26 x 1.5/2 mL Tubes
4 x 50 mL Conical Tubes
8 x 15 mL Conical Tubes
8 x 5 mL Conical Tubes
Unit Size:
Pack of 2
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15108364 8 x 5 mL Conical Tubes Pack of 2
13256589 8 x 15 mL Conical Tubes Pack of 2
13266589 4 x 50 mL Conical Tubes Pack of 2
13276589 1 x 250 mL Tubes Pack of 2
13286589 2 x 50 mL Conical Tubes Pack of 2
12806223 14 x 2.6/8 mL Tubes Pack of 2
13206599 13 x 5.5/12 mL Tubes Pack of 2
13216599 12 x 7/17 mL Tubes Pack of 2
13226599 26 x 1.5/2 mL Tubes Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15108364 Supplier Eppendorf™ Supplier No. 5804793005
Estimated Shipment: 17-12-2026
Available in a variety of configurations to suite a wide range of applications.
  • For use with Centrifuge 5804/5804 R and 5810/5810 R
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Tube Type 5 mL Conical Tube
Capacity 8 x 5 mL Conical Tubes
For Use With (Equipment) S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges