Learn More
Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips
GREENER_CHOICE
Product Code. 12634607 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
25 mL
50 mL
Color Code:
Light Gray
Red
Quantity:
1 Ea.
7 Pk.
Unit Size:
Each
Pack of 7
Product Code. 12634607
Brand: Eppendorf™ 0030089731
Only null left
Add to basket
Add to basket
Use these adapters with Eppendorf pipet tips. Eppendorf™ Adapters for Combitips Advanced™ Pipet Tips are color-coded and autoclavable.
  • Available in 25 and 50mL
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 25 mL
Autoclavable Autoclavable
Quantity 7 Pk.
Color Code Red
For Use With (Equipment) 25 mL Biopur Quality Tip