Learn More
Eppendorf™ twin.tec™ PCR Plates 96-well, 250 μL - PCR Plates
GREENER_CHOICE
Product Code. 10176287 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Colorless
Skirt Style:
Semi-skirted
Unskirted
Unskirted, Divisible
Purity or Quality Grade:
Forensic DNA Grade (Non-sterile)
PCR Clean (Non-sterile)
PCR Clean and Free of Bacterial DNA (Non-sterile)
Packaging:
5 Bags x 4 Plates
Individually Blister Wrapped
Unit Size:
Pack of 10
Pack of 20
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Skirt Style Purity or Quality Grade Packaging unitSize
10176287 Colorless Semi-skirted PCR Clean and Free of Bacterial DNA (Non-sterile) Individually Blister Wrapped Pack of 10
10319597 Blue Semi-skirted PCR Clean and Free of Bacterial DNA (Non-sterile) Individually Blister Wrapped Pack of 10
15665367 Colorless Semi-skirted Forensic DNA Grade (Non-sterile) Individually Blister Wrapped Pack of 10
10608439 Colorless Unskirted PCR Clean (Non-sterile) 5 Bags x 4 Plates Pack of 20
10441528 Blue Unskirted PCR Clean (Non-sterile) 5 Bags x 4 Plates Pack of 20
10297987 Colorless Unskirted, Divisible PCR Clean (Non-sterile) 5 Bags x 4 Plates Pack of 20
10258037 Blue Unskirted, Divisible PCR Clean (Non-sterile) 5 Bags x 4 Plates Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10176287 Supplier Eppendorf™ Supplier No. 0030129326
In Stock
Add to basket
Add to basket
Eppendorf's twin.tec™ PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. With raised well rims, these plates enable an effective seal, minimizing the risk of cross-contamination and contributing to consistently reliable results.
  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Improved well-to-well tolerance.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
Certifications
Certified DNA-, DNase-, RNase-, and PCR-inhibitor-free.
† Certificate, test procedures and detailed information available upon request.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Human DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging Individually Blister Wrapped
Color Colorless
Product Type PCR Plate
Skirt Style Semi-skirted
Material Polypropylene, Polycarbonate
Volume (Metric) Well 250 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) RT-PCR applications
Max. RCF 2250 x g
Purity or Quality Grade PCR Clean and Free of Bacterial DNA (Non-sterile)
Quantity 10 Pk.
Rimmed Yes
Well Color Colorless