Learn More
Eppendorf™ 750mL Round Buckets for S-4-104 Rotor
Product Code. 13296599 Shop All Eppendorf Products
Change view
Click to view available options
Quantity:
2/Pk.
4/Pk.
Unit Size:
Pack of 2
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13296599 2/Pk. Pack of 2
13206609 4/Pk. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13296599 Supplier Eppendorf™ Supplier No. 5820742003
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 09-12-2026
Designed for use with Eppendorf Rotors and Centrifuges
  • Available in packs of two or four
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) S-4-104 rotor for 5810/5810R centrifuge
Capacity 750 mL
Color Blue
Container Type Bucket
Quantity 2/Pk.