missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EphA6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
221.00€ - 305.00€
Specifications
| Antigen | EphA6 |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
EphA6 Polyclonal antibody specifically detects EphA6 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| EphA6 | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 0.2 um filtered solution in PBS | |
| 285220 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, EHK2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, PRO57066 | |
| Produced in rabbits immunized with purified, recombinant Mouse EphA6 (Accession#: NP_031964.2; Met1-Gln546) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title