missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ EpCAM/TROP1 Antibody (oportuzumab) - Humanized, Novus Biologicals™

Human Monoclonal Antibody

213.00€ - 920.00€

Specifications

Antigen EpCAM/TROP1
Clone oportuzumab
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30048035
View Documents
Novus Biologicals™
NBP3-28534-1mg
1 mg
920.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30046825
View Documents
Novus Biologicals™
NBP3-28534-50ug
50 μg
213.00€
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30045130
View Documents
Novus Biologicals™
NBP3-28534-100ug
100 μg
326.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

EpCAM/TROP1
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Cancer, Cancer Stem Cells, Tumor Biomarkers
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
EpCAM/ TROP1/ CD326
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
oportuzumab
Monoclonal
Purified
RUO
Human
P16422
4072
IgG1
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.