missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EpCAM/TROP1 Antibody (CL6313) [DyLight 650], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76486C
This item is not returnable.
View return policy
Description
EpCAM/TROP1 Monoclonal antibody specifically detects EpCAM/TROP1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| EpCAM/TROP1 | |
| Monoclonal | |
| DyLight 650 | |
| 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A | |
| This antibody was developed using a synthetic peptide derived from P16422, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cancer Stem Cells, Tumor Biomarkers | |
| 4072 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL6313 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction