missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 1311 TCLP Pesticide Spiking Mixture 398 2000-4000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Camphechlor (Toxaphene)∣Chlordane (technical) |
| CAS | 8001-35-2, 12789-03-6 |
| Analyte or Component Names | Camphechlor (Toxaphene)∣Chlordane (technical) |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Chlordane (technical) 2000 μg/mL∣Camphechlor (Toxaphene) 4000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction