missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ SealMax (EU-IVD) Filter Tips
Shop All Eppendorf Products

Click to view available options
Volume (Metric):
0.1 to 10 μL
50 to 1000 μL
Length (Metric):
34 mm
76 mm
Unit Size:
Pack of 960
Description
- ep Dualfilter T.I.P.S. SealMax (EU-IVD) filter tips combine the unique and established two-phase filter for protection of both sample and pipette
- Utilizes an innovative liquid and moisture barrier
- Ideally suited for methods where preserving sample integrity and preventing cross-contamination are critical
- Reliable protection from liquids, aerosols, and biomolecules
- No PCR inhibiting additives
- Pipette is protected from aggressive liquids when over-aspiration occurs
- Continuous inspection of each production lot by an independent laboratory guarantees high purity and quality
- Modern slim rack design with reclosable lid
- Improved sustainability with 20% less polypropylene consumed per rack
- Tips are designed and constructed for low-contamination transfer of liquids
- In particular for samples from the human body and for reagents within the scope of an in-vitro diagnostic application in order
- Allows the in-vitro diagnostic device to be used as intended
- ep Dualfilter T.I.P.S.™ SealMax are in-vitro diagnostic devices according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998
- They are intended exclusively for single use and for application by qualified staff
Specifications
Specifications
| Volume (Metric) | 50 to 1000 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Length (Metric) | 76 mm |
| Certifications/Compliance | Directive 98/79/EC |
| Features | PCR Clean |
| Format | Rack |
| Quantity | 960 tips |
| Product Type | Consumables |
| Includes | 10 racks × 96 tips |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction