missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENT1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
204.00€ - 470.00€
Specifications
| Antigen | ENT1 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18300926
|
Novus Biologicals
NBP3-02976-20UL |
20 μL |
204.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18363206
|
Novus Biologicals
NBP3-02976-100UL |
100 μL |
470.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ENT1 Polyclonal specifically detects ENT1 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| ENT1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 2030 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| ENT1es-type, equilibrative nucleoside transporter 1, MGC1465, MGC3778, solute carrier family 29 (nucleoside transporters), member 1, Solute carrier family 29 member 1, solute carrier family 29, member 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ENT1 (NP_001071645.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title