missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENOX2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33360-20ul
This item is not returnable.
View return policy
Description
ENOX2 Monoclonal antibody specifically detects ENOX2 in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ENOX2 | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| COVA1, Cytosolic ovarian carcinoma antigen 1APK1 antigen, ecto-NOX disulfide-thiol exchanger 2, tNOXAPK1, Tumor-associated hydroquinone oxidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 411-610 of human ENOX2 (NP_872114.1).,, Sequence:, AEALKEENDSLRWQLDAYRNEVELLKQEQGKVHREDDPNKEQQLKLLQQALQGMQQHLLKVQEEYKKKEAELEKLKDDKLQVEKMLENLKEKESCASRLCASNQDSEYPLEKTMNSSPIKSEREALLVGIISTFLHVHPFGASIEYICSYLHRLDNKICTSDVECLMGRLQHTFKQEMTGVGASLEKRWKFCGFEGLKLT | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10495 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction