missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENOPH1 Monoclonal Antibody (OTI4A8), TrueMAB™, OriGene
Description
ENOPH1 Monoclonal antibody specifically detects ENOPH1 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ENOPH1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4A8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | RPL27 |
| Gene Accession No. | P61353, P61358 |
| Gene Alias | 2,3-diketo-5-methylthio-1-phosphopentane phosphatase; 2,3-diketo-5-methylthio-1-phosphopentane phosphatase {ECO:0000255; 2310057D15Rik; acireductone synthase; BB183658; C81437; E1; E-1 enzyme; enolase-phosphatase 1; enolase-phosphatase E1; enolase-phosphatase E1 {ECO:0000255; ENOPH1; HAMAP-Rule:MF_03117}; Masa; MASA homolog; MASA homolog {ECO:0000255; MST145; MSTP145; mtnC; RGD1309016 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?