Learn More
ENO3 Antibody (003), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90274PCP
This item is not returnable.
View return policy
Description
ENO3 Monoclonal antibody specifically detects ENO3 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)