missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endophilin 1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68029-1-IG-150UL
This item is not returnable.
View return policy
Description
The Endophilin 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Endophilin 1. This antibody recognizes human, mouse, rat, pig, rabbit antigen. The Endophilin 1 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| Endophilin 1 | |
| Monoclonal | |
| 1000 μg/mL | |
| CNSA2; EEN-B1; FLJ20276; FLJ25015; SH3D2A; SH3P4 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat, Pig, Rabbit | |
| 38-40 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 1G4H1 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 6456 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction