missing translation for 'onlineSavingsMsg'
Learn More

Endoglycan/PODXL2 Antibody (211816) [DyLight 680], Novus Biologicals™

Product Code. 18526856 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18526856 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18526856 Supplier Novus Biologicals Supplier No. FAB1524Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Endoglycan/PODXL2 Monoclonal antibody specifically detects Endoglycan/PODXL2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Endoglycan/PODXL2
Applications Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 211816
Conjugate DyLight 680
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias endoglycan, PODLX2, podocalyxin-like 2, podocalyxin-like protein 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Endoglycan/PODXL2, Gly33-Thr500, Accession # Q9NZ53
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50512
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.