missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Endoglin/CD105 Antibody (MEM-229) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB110-58718
This item is not returnable.
View return policy
Description
Endoglin/CD105 Monoclonal specifically detects Endoglin/CD105 in Human, Porcine, Canine (Negative), Equine (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
| Endoglin/CD105 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 2 to 6 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:200, CyTOF-ready | |
| ENG | |
| recombinant vaccinia virus containing human CD105 (L-isoform) cDNA | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody (clone MEM-229) recognizes CD105 (Endoglin), a 180 kDa type I integral membrane homodimer glycoprotein expressed on vascular endothelial cells (small and large vessels), activated monocytes and tissue macrophages, stromal cells of certain tissues including bone marrow, pre-B lymphocytes in fetal marrow and erythroid precursors in fetal and adult bone marrow; it is also present on syncytiotrophoblast on placenta throughout pregnancy. | |
| Store at 4C. Do not freeze. | |
| IgG2a |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), CyTOF | |
| MEM-229 | |
| Unconjugated | |
| CD105, CD105 antigen, endoglin, ENDOsler-Rendu-Weber syndrome 1, HHT1FLJ41744, ORW, ORW1 | |
| Mouse | |
| 90 kDa | |
| 0.1 mg | |
| Cancer, Cell Biology, Endothelial Cell Markers, Hypoxia, Immunology, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2022 | |
| Human, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction