missing translation for 'onlineSavingsMsg'
Learn More

Enalaprilat, Dr. Ehrenstorfer

Product Code. 30318944
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30318944

Brand: Dr. Ehrenstorfer DREC13119100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Enalaprilat
CAS 76420-72-9
Analyte or Component Names Enalaprilat
Shipping Condition +20°C
Molecular Formula C18 H24 N2 O5
Certifications/Compliance ISO/IEC 17025
Synonym Enalapril Maleate Imp. C (EP), Enalapril Imp. C (EP), (2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic Acid, Enalaprilat, L-Proline, N-[(1S)-1-carboxy-3-phenylpropyl]-L-alanyl-, L-Proline, 1-[N-(1-carboxy-3-phenylpropyl)-L-alanyl]-, (S)-, N-[(1S)-1-Carboxy-3-phenylpropyl]-L-alanyl-L-proline, Enalapril acid, Enalapril diacid, Enalaprilate, Enalaprilic acid, MK 421 diacid, MK 422, N-[1(S)-Carboxy-3-phenylpropyl]-L-alanyl-L-proline, Enalapril Maleate Impurity C, Enalapril Impurity C
InChI Formula InChI=1S/C18H24N2O5/c1-12(16(21)20-11-5-8-15(20)18(24)25)19-14(17(22)23)10-9-13-6-3-2-4-7-13/h2-4,6-7,12,14-15,19H,5,8-11H2,1H3,(H,22,23)(H,24,25)/t12-,14-,15-/m0/s1
SMILES C[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)N2CCC[C@H]2C(=O)O
IUPAC Name (2S)-1-[(2S)-2-[[(1S)-1-carboxy-3-phenylpropyl]amino]propanoyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol) 348.39
Quantity 50 mg
Formula Weight 348.1685 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.