missing translation for 'onlineSavingsMsg'
Learn More

Emylcamate, Mikromol™

Product Code. 30049823
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049823 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049823 Supplier Mikromol Supplier No. MM3364.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Emylcamate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Emylcamate
CAS 78-28-4
Analyte or Component Names Emylcamate
Shipping Condition Room Temperature
Molecular Formula C7 H15 N O2
Certifications/Compliance ISO 17025
Synonym 3-Pentanol, 3-methyl-, 3-carbamate, 3-Pentanol, 3-methyl-, carbamate (8CI,9CI), Carbamic acid, 1-ethyl-1-methylpropyl ester (6CI,7CI), 1-Ethyl-1-methylpropyl carbamate, 3-Methyl-3-pentanol carbamate, 91 JD, Diethyl methyl carbinol urethan, Emylcamate, JD 91, KABI 925, MK 250, Methyl diethyl carbinol urethan, NSC 169882, NSC 169917, Nuncital, Restetal, Statran, Striatran, tert-Hexanol carbamate
InChI Formula InChI=1S/C7H15NO2/c1-4-7(3,5-2)10-6(8)9/h4-5H2,1-3H3,(H2,8,9)
SMILES CCC(C)(CC)OC(=O)N
IUPAC Name 3-methylpentan-3-yl carbamate
Molecular Weight (g/mol) 145.2
Quantity 50 mg
Formula Weight 145.1103 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.