Learn More
Novus Biologicals™ EMMPRIN/CD147 Antibody (Centocor patent anti-CD147) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28146-50ug
 View more versions of this product
Product Code. 30047232
  • 213.00€ / 50µg
Description
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EMMPRIN/CD147
Monoclonal
Unconjugated
Q54A51
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Centocor patent anti-CD147
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
EMMPRIN/ CD147
50 μg
Cancer, Signal Transduction, Vision
682