missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EMILIN1 Antibody (C11A8), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
671.00€
Specifications
| Antigen | EMILIN1 |
|---|---|
| Clone | C11A8 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Conjugate | mFluor Violet 450 SE |
Description
EMILIN1 Monoclonal antibody specifically detects EMILIN1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)Specifications
| EMILIN1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
| mFluor Violet 450 SE | |
| Rat | |
| Mouse | |
| 11117 | |
| IgG2a κ | |
| Protein G purified |
| C11A8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| DKFZp586M121, elastin microfibril interface-located protein 1, elastin microfibril interfacer 1, EMI, EMILIN, EMILIN-1, gp115 | |
| recombinant mouse Emilin-1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title