missing translation for 'onlineSavingsMsg'
Learn More

ELTD1/ETL Antibody (CL4164) [Alexa Fluor™ 750], Novus Biologicals™

Product Code. 30300753 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30300753 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30300753 Supplier Novus Biologicals Supplier No. NBP259041AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ELTD1/ETL Monoclonal antibody specifically detects ELTD1/ETL in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELTD1/ETL
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL4164
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias EGF, latrophilin and seven transmembrane domain containing 1, EGF, latrophilin and seven transmembrane domain-containing protein 1, ETL protein, ETLEGF-TM7-latrophilin-related protein, KPG_003
Host Species Mouse
Immunogen This antibody was developed using a recombinant protein derived from Q9HBW9, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 64123
Target Species Human
Content And Storage Store at 4⪚C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.