missing translation for 'onlineSavingsMsg'
Learn More

ELMO2 Antibody (3G7), Novus Biologicals™

Product Code. 18449469 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18449469 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18449469 Supplier Novus Biologicals Supplier No. H00063916M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ELMO2 Monoclonal antibody specifically detects ELMO2 in Human samples. It is validated for ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELMO2
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3G7
Conjugate Unconjugated
Dilution ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_573403
Gene Alias ced-12 homolog 2, CED12A, ELMO-2, engulfment and cell motility 2, engulfment and cell motility 2 (ced-12 homolog, C. elegans), engulfment and cell motility protein 2, FLJ11656, hCed-12A, KIAA1834CED12CED-12, PH domain protein CED12A, Protein ced-12 homolog A
Host Species Mouse
Immunogen ELMO2 (NP_573403.1, 89 a.a. ∽ 188 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MERTQSSNMETRLDAMKELAKLSADVTFATEFINMDGIIVLTRLVESGTKLLSHYSEMLAFTLTAFLELMDHGIVSWDMVSITFIKQIAGYVSQPMVDVS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 63916
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.