missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELKS Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00€ - 550.00€
Specifications
| Antigen | ELKS |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229483
|
Novus Biologicals
NBP3-38612-100ul |
100 μL |
550.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233132
|
Novus Biologicals
NBP3-38612-20ul |
20 μL |
190.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ELKS Polyclonal antibody specifically detects ELKS in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| ELKS | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ELKS/RAB6-interacting/CAST family member 1, ELKSCAST2, ERC-1, FLJ31750, KIAA1081Cast2, MGC12974, RAB6 interacting protein 2, Rab6-interacting protein 2, RAB6IP2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 700-850 of human ELKS (NP_829884.1).,, Sequence:, EIALEQKKEECLKMESQLKKAHEAALEARASPEMSDRIQHLEREITRYKDESSKAQAEVDRLLEILKEVENEKNDKDKKIAELERQVKDQNKKVANLKHKEQVEKKKSAQMLEEARRREDNLNDSSQQLQDSLRKKDDRIEELEEALRESV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 23085 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title