Learn More
Electrolyte For Lead ISE, Mettler Toledo™
Product Code. 11917721 Shop All Mettler Toledo Products
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11917721 20 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11917721 Supplier Mettler Toledo Supplier No. 51107875
Specifications
Description Electrolyte for Lead ISE, 20mL
Quantity 20 mL
Solution Type Electrolyte
Packaging 20 mL