missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a protein that is one of the two components of elastic fibers. The encoded protein is rich in hydrophobic amino acids such as glycine and proline, which form mobile hydrophobic regions bounded by crosslinks between lysine residues. Deletions and mutations in this gene are associated with supravalvular aortic stenosis (SVAS) and autosomal domit cutis laxa. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Antigen | Elastin |
| Applications | Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.16 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | ELN |
| Gene Accession No. | P15502, P54320 |
| Gene Alias | elastin, ELN, SVAS, Tropoelastin, WBS, WS |
| Gene Symbols | ELN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?