missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5A Antibody (5E1), Novus Biologicals™
Rabbit Monoclonal Antibody
355.00€ - 533.00€
Specifications
| Antigen | eIF5A |
|---|---|
| Clone | 5E1 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043811
|
Novus Biologicals
NBP3-26604-50ul |
50 μL |
355.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043166
|
Novus Biologicals
NBP3-26604-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF5A Monoclonal antibody specifically detects eIF5A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| eIF5A | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| eIF-4D, eIF-5A, eIF-5A1, eIF-5A-1, EIF5A1, eIF5AI, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 1, eukaryotic translation initiation factor 5A, eukaryotic translation initiation factor 5A-1, MGC104255, MGC99547, Rev-binding factor | |
| A synthesized peptide derived from Human eIF5A [UniProt P63241] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 5E1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1984 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title