missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ EIF4G3 Recombinant Protein

Product Code. 16192752
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16192752

Brand: Abnova™ H00008672P01.10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH30578
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 8672
Molecular Weight (g/mol) 82.39
Name EIF4G3 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MNSQPQTRSPFAAGPRPPHHQFFQRPQIQPPRATIPNSSPSIRPGAQTPTAVYQANQHIMMVNHLPMPYPVPQGPQYCIPQYRHSGPPYVGPPQQYPVQPPGPGPFYPGPGPGDFPNAYGTPFYPSQPVYQSAPIMVPTQQQPPPAKREKKTIRIRDPNQGGKDITEEIISGGGSRNPTPPIGRPTSTPTPPQQLPSQVPEHSPVVYGTVESAHLAASTPVTAASDQKQEEKPKPDPVLKSPSPVLRLVLSGEKKEQEGQTSETTAIVSIAELPLPPSPTTVSSVARSTIAAPTSSALSSQPIFTTAIDDRCELSSPREDTIPIPSLTSCTETSDPLPTNENDGDICKKPCSVAPNDIPLVSSTNLINEINGVSEKLSATESIVEIVKQEVLPLTLELEILENPPEEMKLECIPAPITPSTVPSFPPTPPTPPASPPHTPVIVPAAATTVSSPSAAITVQRVLEEDESIRTCLSEDAKEIQNKIEVEADGQTEEILDSQNLNSRRSPVPETSNEC
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias eIF4GII
Common Name EIF4G3
Gene Symbol EIF4G3
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.