missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2S1, Mouse anti-Human, Clone: 3H12-C11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00001965-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MPGLSCRFYQHKFPEVEDVVMVNVRSIAEMGAYVSLLEYNNIEGMILLSELSRRRIRSINKLIRIGRNECVVVIRVDKEKGYIDLSKRRVSPEEAIKCEDKFTKSKTVYSILRHVAEVLEYTKDEQLESLFQRTAWVFDDKYKRPGYGAYDAFKHAVSDPSILDSLDLNEDEREVLINNINRRLTPQAVKIRADIEVACYGYEGIDAVKEALRAGLNCSTENMPIKINLIAPPRYVMTTTTLERTEGLSVLSQAMAVIKEKIEEKRGVFNVQMEPKVVTDTDETELARQMERLERENAEVDGDDDAEEMEAKAEDSpecifications
| EIF2S1 | |
| Monoclonal | |
| Unconjugated | |
| eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa | |
| EIF-2/EIF-2A/EIF-2alpha/EIF2/EIF2A | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, KnockDown, Western Blot | |
| 3H12-C11 | |
| In 1x PBS, pH 7.4 | |
| BC002513 | |
| EIF2S1 | |
| EIF2S1 (AAH02513.1, 1 a.a. ∼ 315 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 1965 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction