missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2A Rabbit anti-Human, Mouse, Rat, Clone: 8V8H7, Novus Biologicals™
Rabbit Monoclonal Antibody
214.00€ - 508.00€
Specifications
| Antigen | eIF2A |
|---|---|
| Clone | 8V8H7 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18392505
|
Novus Biologicals
NBP3-16864-20UL |
20 μg |
214.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18325694
|
Novus Biologicals
NBP3-16864-100UL |
100 μg |
508.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
eIF2A Monoclonal antibody specifically detects eIF2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Tekniske data
| eIF2A | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EIF-2, EIF-2A, EIF2AEIF2, EIF-2alpha, eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa, subunit 1 (alpha, 35kD ) | |
| A synthetic peptide corresponding to a sequence within amino acids 450-550 of human eIF2A (Q9BY44). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 8V8H7 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, DNA Repair, Lipid and Metabolism, Translation Control, Unfolded Protein Response | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 83939 | |
| IgG | |
| Affinity purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel