missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF2A Antibody (3A7B11), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-26296DL594
This item is not returnable.
View return policy
Description
eIF2A Monoclonal specifically detects eIF2A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| eIF2A | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EIF2A | |
| Partial recombinant human EIF2A (between residues 350-585) expressed in E. coli [Uniprot Q9BY44] | |
| 0.1 mL | |
| Chromatin Research, DNA Repair, Lipid and Metabolism, Phospho Specific, Translation Control, Unfolded Protein Response | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3A7B11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| EIF-2, EIF-2A, EIF2AEIF2, EIF-2alpha, eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa, subunit 1 (alpha, 35kD ) | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction