missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EID1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47355
This item is not returnable.
View return policy
Description
EID1 Polyclonal specifically detects EID1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| EID1 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| 21 kDa pRb-associated protein, C15orf3PTD014, CREBBP/EP300 inhibitor 1, CREBBP/EP300 inhibitory protein 1MGC138884, CRI1EP300-interacting inhibitor of differentiation 1, E1A-like inhibitor of differentiation 1, EID-1MGC138883, EP300 interacting inhibitor of differentiation 1, IRO45620, NB4 apoptosis related protein, Rb- and p300-binding protein EID-1, RBP21, retinoblastoma protein-associated protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| EID1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MSEMAELSELYEESSDLQMDVMPGEGDLPQMEVGSGSRELSLRPSRSGAQQLEEEGPMEEEEAQPMAAPEGKRSLANGPNAGEQPGQVAGADFESED | |
| 0.1 mL | |
| Vision | |
| 23741 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction